Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072344.3 | NP_504745.1 | 6 - 7 | 732 | 130 | |
GenBank mRNA | AY008132.1 | - | 6 - 7 | 724 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |