Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028881.4 | NP_001024052.1 | 3 - 4 | 233 | 70 | |
NM_001028882.5 | NP_001024053.2 | 3 - 4 | 233 | 70 | ||
NM_001269144.3 | NP_001256073.1 | 2 - 3 | 449 | 70 | ||
NM_001307743.4 | NP_001294672.1 | 2 - 3 | 181 | 70 | ||
GenBank mRNA | AF166169.1 | - | 2 - 3 | 181 | 70 | |
AF166170.1 | - | 2 - 3 | 181 | 70 | ||
AF261891.1 | - | 3 - 4 | 249 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |