Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028881.4 | NP_001024052.1 | 4 - 5 | 314 | 76 | |
NM_001028882.5 | NP_001024053.2 | 4 - 5 | 314 | 76 | ||
NM_001269144.3 | NP_001256073.1 | 3 - 4 | 530 | 76 | ||
NM_001307743.4 | NP_001294672.1 | 3 - 4 | 262 | 76 | ||
GenBank mRNA | AF166169.1 | - | 3 - 4 | 262 | 76 | |
AF166170.1 | - | 3 - 4 | 262 | 76 | ||
AF261891.1 | - | 4 - 5 | 330 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |