Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028882.5 | NP_001024053.2 | 8 - 9 | 6391 | 66 | |
GenBank mRNA | AF166169.1 | - | 7 - 8 | 6339 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |