Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072410.7 | NP_504811.2 | 3 - 4 | 575 | 166 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |