Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072413.7 | NP_504814.1 | 1 - 2 | 136 | 73 | |
GenBank mRNA | AF260243.1 | - | 1 - 2 | 121 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |