Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269154.3 | NP_001256083.1 | 3 - 4 | 541 | 93 | |
NM_001269155.2 | NP_001256084.1 | 4 - 5 | 641 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |