Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072479.6 | NP_504880.2 | 6 - 7 | 1422 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |