Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028646.1 | NP_001023817.1 | 5 - 6 | 967 | 84 | |
NM_001028647.2 | NP_001023818.1 | 6 - 7 | 1090 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |