Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072636.2 | NP_505037.1 | 11 - 12 | 1299 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |