Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072641.4 | NP_505042.3 | 4 - 5 | 855 | 171 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |