Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072663.6 | NP_505064.1 | 3 - 4 | 400 | 72 | |
NM_072664.5 | NP_505065.1 | 2 - 3 | 293 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |