Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072673.7 | NP_505074.1 | 1 - 2 | 210 | 68 | |
GenBank mRNA | AB072926.1 | - | 1 - 2 | 192 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |