Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072693.4 | NP_505094.1 | 11 - 12 | 5669 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |