Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072693.4 | NP_505094.1 | 7 - 8 | 1404 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |