Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028652.7 | NP_001023823.1 | 2 - 3 | 389 | 108 | |
NM_001028653.5 | NP_001023824.1 | 1 - 2 | 281 | 108 | ||
NM_001129452.4 | NP_001122924.1 | 2 - 3 | 302 | 108 | ||
NM_001383352.1 | 2 - 3 | 384 | 108 | |||
GenBank mRNA | AY525083.1 | - | 1 - 2 | 281 | 108 | |
AY525084.1 | - | 1 - 2 | 281 | 108 | ||
AY525085.1 | - | 1 - 2 | 281 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |