Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129452.4 | NP_001122924.1 | 5 - 6 | 954 | 65 | |
NM_001383352.1 | 5 - 6 | 1036 | 65 | |||
GenBank mRNA | AY525083.1 | - | 4 - 5 | 933 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |