Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072753.6 | NP_505154.1 | 4 - 5 | 537 | 69 | |
GenBank mRNA | AF013952.1 | - | 4 - 5 | 501 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |