Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072756.5 | NP_505157.1 | 2 - 3 | 223 | 105 | |
GenBank mRNA | AY008125.1 | - | 2 - 3 | 214 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |