Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072787.4 | NP_505188.3 | 16 - 17 | 3211 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |