Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028738.7 | NP_001023909.2 | 2 - 3 | 515 | 89 | |
NR_138495.1 | 2 - 3 | 513 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |