Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072827.3 | NP_505228.2 | 3 - 4 | 406 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |