Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072912.7 | NP_505313.1 | 15 - 16 | 2423 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |