Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072940.5 | NP_505341.1 | 7 - 8 | 815 | 82 | |
NR_145827.1 | 5 - 6 | 549 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |