Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072940.5 | NP_505341.1 | 11 - 12 | 1618 | 96 | |
NR_145827.1 | 9 - 10 | 1352 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |