Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028658.4 | NP_001023829.1 | 2 - 3 | 699 | 78 | |
NM_001028659.5 | NP_001023830.1 | 2 - 3 | 660 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |