Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322581.3 | NP_001309624.1 | 3 - 4 | 598 | 98 | |
NM_072977.3 | NP_505378.1 | 2 - 3 | 739 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |