Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028968.4 | NP_001024139.1 | 2 - 3 | 459 | 87 | |
NM_001028969.6 | NP_001024140.1 | 2 - 3 | 465 | 87 | ||
NM_001373070.2 | NP_001360570.1 | 2 - 3 | 469 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |