Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072994.2 | NP_505395.2 | 5 - 6 | 769 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |