Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073004.1 | NP_505405.1 | 1 - 2 | 83 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |