Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322582.2 | NP_001309498.1 | 4 - 5 | 745 | 161 | |
NR_136369.1 | 4 - 5 | 721 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |