Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330875.4 | NP_001317742.1 | 12 - 13 | 4165 | 74 | |
NM_001330876.2 | NP_001317741.1 | 12 - 13 | 4159 | 74 | ||
NM_001330877.3 | NP_001317812.1 | 6 - 7 | 1176 | 74 | ||
NM_001330878.3 | NP_001317813.1 | 6 - 7 | 1170 | 74 | ||
NM_001330879.3 | NP_001317814.1 | 5 - 6 | 1059 | 74 | ||
NM_001330880.3 | NP_001317815.1 | 5 - 6 | 1053 | 74 | ||
NM_001330881.3 | NP_001317816.1 | 4 - 5 | 1011 | 74 | ||
NM_001373073.2 | NP_001359965.1 | 13 - 14 | 4086 | 74 | ||
NM_001380727.2 | NP_001367720.1 | 12 - 13 | 4159 | 74 | ||
NM_171505.6 | NP_741590.1 | 2 - 3 | 254 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |