Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hum-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330875.4 | NP_001317742.1 | 14 - 15 | 4382 | 97 | |
NM_001330876.2 | NP_001317741.1 | 14 - 15 | 4376 | 97 | ||
NM_001330877.3 | NP_001317812.1 | 8 - 9 | 1393 | 97 | ||
NM_001330878.3 | NP_001317813.1 | 8 - 9 | 1387 | 97 | ||
NM_001330879.3 | NP_001317814.1 | 7 - 8 | 1276 | 97 | ||
NM_001330880.3 | NP_001317815.1 | 7 - 8 | 1270 | 97 | ||
NM_001330881.3 | NP_001317816.1 | 6 - 7 | 1228 | 97 | ||
NM_001373073.2 | NP_001359965.1 | 15 - 16 | 4303 | 97 | ||
NM_171505.6 | NP_741590.1 | 4 - 5 | 471 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |