Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgef-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276913.2 | NP_001263842.1 | 1 - 2 | 153 | 162 | |
NM_073068.4 | NP_505469.3 | 1 - 2 | 150 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |