Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028876.3 | NP_001024047.1 | 12 - 13 | 1825 | 76 | |
NM_001269197.4 | NP_001256126.1 | 13 - 14 | 2263 | 76 | ||
NM_001269198.4 | NP_001256127.1 | 13 - 14 | 1913 | 76 | ||
NM_001373076.2 | NP_001360549.1 | 13 | 2224 | 76 | ||
NM_001373077.2 | NP_001360548.1 | 13 | 1879 | 76 | ||
NM_001373078.2 | NP_001360550.1 | 12 - 13 | 1808 | 76 | ||
GenBank mRNA | AB125702.1 | - | 12 - 13 | 1847 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |