Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073088.4 | NP_505489.1 | 6 - 7 | 3241 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |