Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029139.3 | NP_001024310.1 | 1 - 2 | 261 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |