Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sru-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269214.1 | NP_001256143.1 | 5 - 6 | 797 | 146 | |
NM_001269215.2 | NP_001256144.1 | 4 - 5 | 603 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |