Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsy-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269216.3 | NP_001256145.1 | 10 - 11 | 2358 | 93 | |
NM_001269217.3 | NP_001256146.1 | 8 - 9 | 1496 | 93 | ||
NM_001269218.3 | NP_001256147.1 | 11 - 12 | 2483 | 93 | ||
NM_001276918.3 | NP_001263847.1 | 10 - 11 | 2348 | 93 | ||
NM_001276919.3 | NP_001263848.1 | 8 - 9 | 1496 | 93 | ||
NM_001276920.4 | NP_001263849.1 | 11 - 12 | 2483 | 93 | ||
NM_001373080.3 | NP_001359747.1 | 9 | 1717 | 93 | ||
NM_001373081.2 | NP_001360180.1 | 9 | 1698 | 93 | ||
NM_001373082.2 | NP_001359576.1 | 9 | 1717 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |