Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073156.1 | NP_505557.1 | 3 - 4 | 562 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |