Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269228.3 | NP_001256157.1 | 10 - 11 | 4818 | 76 | |
NM_001269229.3 | NP_001256158.1 | 8 - 9 | 3960 | 76 | ||
NM_001269230.3 | NP_001256159.1 | 6 - 7 | 3687 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |