Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ruvb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073166.5 | NP_505567.2 | 4 - 5 | 1442 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |