Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lea-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269231.3 | NP_001256160.1 | 6 - 7 | 2046 | 77 | |
NM_001269233.4 | NP_001256162.1 | 6 - 7 | 2046 | 77 | ||
NM_001269234.3 | NP_001256163.1 | 5 - 6 | 1998 | 77 | ||
NM_001269236.3 | NP_001256165.1 | 5 - 6 | 1998 | 77 | ||
NM_001269240.3 | NP_001256169.1 | 4 - 5 | 1782 | 77 | ||
NM_001269242.2 | NP_001256171.1 | 4 - 5 | 1800 | 77 | ||
NM_001373085.2 | NP_001360513.1 | 6 | 2046 | 77 | ||
NM_001373087.2 | NP_001360515.1 | 5 | 1998 | 77 | ||
NM_001373089.2 | NP_001360517.1 | 4 | 1782 | 77 | ||
GenBank mRNA | AF016513.1 | - | 4 - 5 | 1814 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |