Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129369.6 | NP_001122841.1 | 1 - 2 | 218 | 181 | |
NM_073196.7 | NP_505597.2 | 1 - 2 | 200 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |