Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129369.6 | NP_001122841.1 | 3 - 4 | 535 | 130 | |
NM_073196.7 | NP_505597.2 | 3 - 4 | 517 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |