Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073208.5 | NP_505609.2 | 6 - 7 | 1058 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |