Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073210.3 | NP_505611.2 | 2 - 3 | 269 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |