Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269275.3 | NP_001256204.1 | 15 - 16 | 2554 | 138 | |
NM_001269276.3 | NP_001256205.1 | 6 - 7 | 939 | 138 | ||
NM_001269277.3 | NP_001256206.1 | 4 - 5 | 482 | 138 | ||
NM_001307803.4 | NP_001294732.1 | 4 - 5 | 522 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |