Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307767.1 | NP_001294696.1 | 16 - 17 | 2983 | 89 | |
NM_073249.3 | NP_505650.3 | 16 - 17 | 2971 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |