Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373099.3 | NP_001360575.1 | 6 - 7 | 998 | 132 | |
NM_073321.6 | NP_505722.1 | 6 - 7 | 1000 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |